Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POLDIP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA127942
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA127942 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA127942 Supplier Invitrogen™ Supplier No. PA127942
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

By Western blot, this antibody has only been tested on fusion protein.

POLDIP2 is a protein that interacts with the DNA polymerase delta p50 subunit, as well as with proliferating cell nuclear antigen. The encoded protein maybe play a role in the ability of the replication fork to bypass DNA lesions. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POLDIP2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Poldip2
Gene Accession No. Q91VA6, Q9Y2S7
Gene Alias 1300003F06Rik; 38 kDa DNA polymerase delta interaction protein; DKFZp586F1524; DNA polymerase delta interacting protein 2; DNA-directed polymerase delta interacting protein 2; HSPC017; mitogenin 1; mitogenin I; p38; PDIP38; POLD4; POLDIP2; polymerase (DNA) delta interacting protein 2; polymerase (DNA-directed), delta interacting protein 2; polymerase delta interacting protein 38; polymerase delta-interacting protein 2
Gene Symbols Poldip2
Host Species Chicken
Immunogen Synthetic peptide - human Polymerase delta-interacting protein 2.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26073, 287544, 67811
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgY
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.