Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ POLDIP3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA582625
Description
Immunogen sequence: SLDELIRKRG AAAKGRLNAR PGVGGVRSRV GIQQGLLSQS TRTATFQQRF DARQKIGLSD ARLKLGVKDA REKLLQKDAR FRIKGKVQDA R.
This gene encodes a protein that interacts with the DNA polymerase delta p50 subunit. This protein is a specific target of S6 kinase 1 and regulates cell growth. Two transcript variants that encode different protein isoforms have been identified.
Specifications
POLDIP3 | |
Polyclonal | |
Unconjugated | |
POLDIP3 | |
1110008P04Rik; 46 kDa DNA polymerase delta interaction protein; AA408269; AL022852; C77954; DNA polymerase delta interacting protein 3; KIAA1649; mKIAA1649; p46; PDIP46; POLDIP3; polymerase (DNA) delta interacting protein 3; polymerase (DNA-directed), delta interacting protein 3; polymerase delta interacting protein 46; polymerase delta-interacting protein 3; RNA-binding protein P46; S6K1 Aly/REF-like target; SKAR | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
315170, 73826, 84271 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8BG81, Q9BY77 | |
POLDIP3 | |
Recombinant protein corresponding to Human POLDIP3. Recombinant protein control fragment (Product #RP-92352). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction