Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POLDIP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531966
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531966 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531966 Supplier Invitrogen™ Supplier No. PA531966
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: NT2D1, IMR32, U87-MG, MCF-7. Predicted reactivity: Mouse (90%), Rat (89%), Rhesus Monkey (98%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a protein that interacts with the DNA polymerase delta p50 subunit. This protein is a specific target of S6 kinase 1 and regulates cell growth. Two transcript variants that encode different protein isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POLDIP3
Applications Western Blot
Classification Polyclonal
Concentration 0.63 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene POLDIP3
Gene Accession No. Q9BY77
Gene Alias 1110008P04Rik; 46 kDa DNA polymerase delta interaction protein; AA408269; AL022852; C77954; DNA polymerase delta interacting protein 3; KIAA1649; mKIAA1649; p46; PDIP46; POLDIP3; polymerase (DNA) delta interacting protein 3; polymerase (DNA-directed), delta interacting protein 3; polymerase delta interacting protein 46; polymerase delta-interacting protein 3; RNA-binding protein P46; S6K1 Aly/REF-like target; SKAR
Gene Symbols POLDIP3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human POLDIP3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84271
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.