Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POLE Monoclonal Antibody (93H3A), Invitrogen™

Catalog No. PIMA513616
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA513616 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA513616 Supplier Thermo Scientific Supplier No. MA513616
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-13616 targets DNA Polymerase epsilon in IP and WB applications and shows reactivity with Bovine, Human, mouse, and Non-human primate samples. The MA5-13616 immunogen is purified recombinant human DNA polymerase epsilon corresponding to aa1-176.

POLE (Polepsi) is present in proliferating cells and is implicated in repair of UV-damaged DNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POLE
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 93H3A
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Gene POLE
Gene Accession No. Q07864, Q9WVF7
Gene Alias CRCS12; DNA polymerase epsilon catalytic subunit A; DNA polymerase epsilon catalytic subunit protein; DNA polymerase epsilon, catalytic subunit; DNA polymerase II subunit A; DNA-directed DNA polymerase epsilon; FILS; Pole; POLE1; pol-epsilon; polymerase (DNA directed), epsilon; polymerase (DNA directed), epsilon, catalytic subunit; polymerase (DNA) epsilon, catalytic subunit
Gene Symbols POLE
Host Species Mouse
Immunogen Purified recombinant human DNA polymerase epsilon corresponding to aa1-176
Purification Method Protein G
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18973, 5426, 785457
Target Species Bovine, Human, Mouse, Non-human Primate
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.