Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POLE Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584367
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584367 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584367 Supplier Invitrogen™ Supplier No. PA584367
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HLQRHNHLLW LSPTARPDLG GKEADDNCLV MEFDDQATVE INSSGCYSTV CVELDLQNLA VNTILQSHHV NDMEGADSMG ISFDVIQ Highest antigen sequence indentity to the following orthologs: Rat - 93%, Mouse - 93%.

POLE (Polepsi) is present in proliferating cells and is implicated in repair of UV-damaged DNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POLE
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene POLE
Gene Accession No. Q07864
Gene Alias CRCS12; DNA polymerase epsilon catalytic subunit A; DNA polymerase epsilon catalytic subunit protein; DNA polymerase epsilon, catalytic subunit; DNA polymerase II subunit A; DNA-directed DNA polymerase epsilon; FILS; Pole; POLE1; pol-epsilon; polymerase (DNA directed), epsilon; polymerase (DNA directed), epsilon, catalytic subunit; polymerase (DNA) epsilon, catalytic subunit
Gene Symbols POLE
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human POLE. Recombinant protein control fragment (Product #RP-106829).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5426
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.