Learn More
                                    Invitrogen™ POLE2 Polyclonal Antibody
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110195
Description
Immunogen sequence: MAPERLRSRA LSAFKLRGLL LRGEAIKYLT EALQSISELE LEDKLEKIIN AVEKQPLSSN MIERSVVEAA VQECSQSVDE TIEHVFNIIG AFDIPRFVYN SERKKFLPLL MTNHPAPNLF GTPRDKAEMF RERYTILHQR THRHELFTPP VIGSHPDESG SKFQLKTIET LLGSTTKIGD AIVLGMITQL KEGKFFLEDP TGTVQLDLSK AQFHSGLYTE ACFVLAEGWF EDQVFHVNAF GFPPTEPSST TRAYYGNINF FGGPSNTSVK TSAKLKQLEE ENKDAMFVFL.
DNA replication, recombination and repair, all of which are necessary for genome stability, require the presence of exonucleases. In DNA replication, these enzymes are involved in the processing of Okazaki fragments, whereas in DNA repair, they function to excise damaged DNA fragments and correct recombinational mismatches. Exonucleases involved in these processes include DNA polymerases, including DNA pol delta and epsilon. DNA pol delta consists of two subunits-p125 which interacts directly with the sliding DNA clamp protein PCNA, and p50. DNA pol delta can be regulated by cell cycle proteins. DNA pol epsilon is a multiple subunit enzyme, the catalytic subunit of which is encoded by the POL2 gene. The exact reactions catalyzed by DNA pol delta and epsilon on leading and lagging strands have not yet been elucidated. 
            Specifications
| POLE2 | |
| Polyclonal | |
| Unconjugated | |
| POLE2 | |
| DNA pol epsilon2; DNA pol epsilonB; DNA polymerase epsilon 2, accessory subunit; DNA polymerase epsilon small subunit; DNA polymerase epsilon subunit 2; DNA polymerase epsilon subunit B; DNA polymerase epsilon, subunit 2; DNA polymerase II subunit 2; DNApol epsilon2; DNApol epsilonB; DPE2; POLE2; polymerase (DNA directed), epsilon 2 (p59 subunit); polymerase (DNA directed), epsilon 2, accessory subunit; polymerase (DNA) epsilon 2, accessory subunit | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 18974, 5427 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | 
| ELISA, Western Blot | |
| 0.74 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| O54956, P56282 | |
| POLE2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-290 of human POLE2 (NP_002683.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG | 
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
