Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ POLE2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA536952
Description
This antibody detects endogenous protein at a molecular weight of 60 kDa. Purity is >95% by SDS-PAGE.
DNA replication, recombination and repair, all of which are necessary for genome stability, require the presence of exonucleases. In DNA replication, these enzymes are involved in the processing of Okazaki fragments, whereas in DNA repair, they function to excise damaged DNA fragments and correct recombinational mismatches. Exonucleases involved in these processes include DNA polymerases, including DNA pol delta and epsilon. DNA pol delta consists of two subunits-p125 which interacts directly with the sliding DNA clamp protein PCNA, and p50. DNA pol delta can be regulated by cell cycle proteins. DNA pol epsilon is a multiple subunit enzyme, the catalytic subunit of which is encoded by the POL2 gene. The exact reactions catalyzed by DNA pol delta and epsilon on leading and lagging strands have not yet been elucidated.
Specifications
POLE2 | |
Polyclonal | |
Unconjugated | |
POLE2 | |
DNA pol epsilon2; DNA pol epsilonB; DNA polymerase epsilon 2, accessory subunit; DNA polymerase epsilon small subunit; DNA polymerase epsilon subunit 2; DNA polymerase epsilon subunit B; DNA polymerase epsilon, subunit 2; DNA polymerase II subunit 2; DNApol epsilon2; DNApol epsilonB; DPE2; POLE2; polymerase (DNA directed), epsilon 2 (p59 subunit); polymerase (DNA directed), epsilon 2, accessory subunit; polymerase (DNA) epsilon 2, accessory subunit | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18974, 299112, 5427 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
O54956, P56282 | |
POLE2 | |
Synthetic peptide corresponding to amino acids 144-189 of human DNA pol epsilon 2. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction