Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PolE3/p17 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501571759
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-1759 100 μL
50-157-1760 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-157-1759 Supplier Bethyl Laboratories, Inc Supplier No. IHC00250
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat, Bovine, Guinea pig_10141, Panda, Orangutan, Monkey, Gorilla, Chimpanzee, Northern whi

POLE3 is a histone-fold protein that interacts with other histone-fold proteins to bind DNA in a sequence-independent manner. These histone-fold protein dimers combine within larger enzymatic complexes for DNA transcription, replication, and packaging.

Specifications

Antigen PolE3/p17
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene POLE3
Gene Accession No. Q9JKP7, Q9NRF9
Gene Alias CHRAC 17, Chromatin Accessibility Complex 17, DNA Polymerase Epsilon p17 Subunit, DNA Polymerase Epsilon Subunit 3, 17, YBL1
Gene Symbols POLE3
Host Species Rabbit
Immunogen Between 97 and 147
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54107, 59001
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.