Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POLE3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA592999
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA592999 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA592999 Supplier Invitrogen™ Supplier No. PA592999
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAERPEDLNL PNAVITRIIK EALPDGVNIS KEARSAISRA ASVFVLYATS CANNFAMKGK RKTLNASDVL SAMEEMEFQR FVTPLKEALE AYRREQKGKK EASEQKKKDK DKKTDSEEQD KSRDEDNDED EERLEEEEQN EEEEVDN; Positive Samples: HepG2, Mouse testis, Mouse bone marrow, Mouse spleen; Cellular Location: Nucleus.

Mitochondrial creatine kinase (MtCK) is responsible for the transfer of high energy phosphate from mitochondria to the cytosolic carrier, creatine. It belongs to the creatine kinase isoenzyme family. It exists as two isoenzymes, sarcomeric MtCK and ubiquitous MtCK, encoded by separate genes. Mitochondrial creatine kinase occurs in two different oligomeric forms: dimers and octamers, in contrast to the exclusively dimeric cytosolic creatine kinase isoenzymes. Sarcomeric mitochondrial creatine kinase has 80% homology with the coding exons of ubiquitous mitochondrial creatine kinase. This gene contains sequences homologous to several motifs that are shared among some nuclear genes encoding mitochondrial proteins and thus may be essential for the coordinated activation of these genes during mitochondrial biogenesis. Three transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POLE3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.44 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene POLE3
Gene Accession No. Q9JKP7, Q9NRF9
Gene Alias 1810034K18Rik; arsenic transactivated protein; arsenic-transactivated protein; AsTP; CHARAC17; CHRAC 17; CHRAC17; CHRAC-17; Chromatin accessibility complex 17 kDa protein; DNA polymerase epsilon 3, accessory subunit; DNA polymerase epsilon p17 subunit; DNA polymerase epsilon subunit 3; DNA polymerase epsilon subunit p17; DNA polymerase epsilon, subunit 3; DNA polymerase II subunit 3; DNA-directed DNA polymerase epsilon 3; histone fold protein CHRAC17; huCHRAC17; NF-YB-like protein; p17; Pole3; polymerase (DNA directed), epsilon 3 (p17 subunit); polymerase (DNA directed), epsilon 3, accessory subunit; polymerase (DNA) epsilon 3, accessory subunit; YBL1; YB-like protein 1
Gene Symbols POLE3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-147 of human POLE3 (NP_0591393).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54107, 59001
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.