Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ POLG Recombinant Rabbit Monoclonal Antibody (9V9D9)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA537845
Description
Positive Samples: HT-29, 293T, Mouse testis, Mouse brain, Rat liver, Rat testis Immunogen sequence: LVREEDRYRA ALALQITNLL TRCMFAYKLG LNDLPQSVAF FSAVDIDRCL RKEVTMDCKT PSNPTGMERR YGIPQGEALD IYQIIELTKG SLEKRSQPGP.
Mitochondrial DNA polymerase is heterotrimeric, consisting of a homodimer of accessory subunits plus a catalytic subunit. The protein encoded by this gene is the catalytic subunit of mitochondrial DNA polymerase. The encoded protein contains a polyglutamine tract near its N-terminus that may be polymorphic. Defects in this gene are a cause of progressive external ophthalmoplegia with mitochondrial DNA deletions 1 (PEOA1), sensory ataxic neuropathy dysarthria and ophthalmoparesis (SANDO), Alpers-Huttenlocher syndrome (AHS), and mitochondrial neurogastrointestinal encephalopathy syndrome (MNGIE). Two transcript variants encoding the same protein have been found for this gene.
Specifications
POLG | |
Recombinant Monoclonal | |
0.8 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P54098, P54099, Q9QYV8 | |
POLG | |
A synthetic peptide corresponding to a sequence within amino acids 1140-1239 of human DNA Polymerase gamma (P54098). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Western Blot | |
9V9D9 | |
Unconjugated | |
POLG | |
AA409516; DNA polymerase gamma; DNA polymerase gamma subunit 1 (Mitochondrial DNA polymerase catalytic subunit) (PolG-alpha); DNA polymerase gamma, catalytic subunit; DNA polymerase subunit gamma-1; DNA-directed DNA polymerase gamma; FLJ27114; MDP1; Mip1; MIRAS; Mitochondrial DNA polymerase catalytic subunit; mitochondrial DNA polymerase gamma; mitochondrial DNA polymerase gamma catalytic subunit; mitochondrial DNA polymerase-gamma; mitochondrial polymerase gamma; MTDPS4A; MTDPS4B; PEO; Pol gamma; Polg; POLG1; POLGA; PolG-alpha; polymerase (DNA directed), gamma; polymerase (DNA) gamma, catalytic subunit; polymerase, gamma; SANDO; SCAE; truncated mitochondrial DNA polymerase gamma catalytic subunit | |
Rabbit | |
Affinity Chromatography | |
RUO | |
18975, 5428, 85472 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction