Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POLG2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110196
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110196 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110196 Supplier Invitrogen™ Supplier No. PA5110196
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HPVFDTKQIR NGVKSIGEKT EASLVWFTPP RTSNQWLDFW LRHRLQWWRK FAMSPSNFSS SDCQDEEGRK GNKLYYNFPW GKELIETLWN LGDHELLHMY PGNVSKLHGR DGRKNVVPCV LSVNGDLDRG MLAYLYDSFQ LTENSFTRKK NLHRKVLKLH PCLAPIKVAL DVGRGPTLEL RQVCQGLFNE LLENGISVWP GYLETMQSSL EQLYSKYDEM SILFTVLVTE TTLENGLIHL RSRDTTMKEM MHISKLKDFL IKYISSAKNV.

This gene encodes the processivity subunit of the mitochondrial DNA polymerase gamma. The encoded protein forms a heterotrimer containing one catalytic subunit and two processivity subunits. This protein enhances DNA binding and promotes processive DNA synthesis. Mutations in this gene result in autosomal dominant progressive external ophthalmoplegia with mitochondrial DNA deletions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POLG2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.02 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene POLG2
Gene Accession No. Q9QZM2, Q9UHN1
Gene Alias AA409516; DNA polymerase gamma; DNA polymerase gamma 2, accessory subunit; DNA polymerase gamma accessory 55 kDa subunit; DNA polymerase gamma subunit 1 (Mitochondrial DNA polymerase catalytic subunit) (PolG-alpha); DNA polymerase gamma, catalytic subunit; DNA polymerase subunit gamma-1; DNA polymerase subunit gamma-2, mitochondrial; DNA-directed DNA polymerase gamma; HP55; MDP1; Mip1; MIRAS; Mitochondrial DNA polymerase accessory subunit; mitochondrial DNA polymerase catalytic subunit; mitochondrial DNA polymerase gamma; mitochondrial DNA polymerase gamma catalytic subunit; mitochondrial DNA polymerase subunit gamma-2; mitochondrial DNA polymerase, accessory subunit; mitochondrial DNA polymerase-gamma; mitochondrial polymerase gamma; mitochondrial polymerase gamma 2; MTDPS4A; MTDPS4B; MTPOLB; p55; PEO; PEOA4; Pol gamma; POLB; POLG; POLG1; Polg2; POLGA; PolG-alpha; POLGB; polG-beta; polymerase (DNA directed), gamma; polymerase (DNA directed), gamma 2, accessory subunit; polymerase (DNA) gamma 2, accessory subunit; polymerase (DNA) gamma, catalytic subunit; polymerase, gamma; polymerase, gamma 2; SANDO; SCAE; truncated mitochondrial DNA polymerase gamma catalytic subunit
Gene Symbols POLG2
Host Species Rabbit
Immunogen Recombinant protein (or fragment)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11232, 303612, 50776
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.