Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ POLI Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA316539
Description
Suggested positive control: HeLa whole cell extract (100 μg) or GM0193 (lymphoblastoid cell line)..
Error-prone DNA polymerase specifically involved in DNA repair. Plays an important role in translesion synthesis, where the normal high-fidelity DNA polymerases cannot proceed and DNA synthesis stalls. Favors Hoogsteen base-pairing in the active site. Inserts the correct base with high-fidelity opposite an adenosine template. Exhibits low fidelity and efficiency opposite a thymidine template, where it will preferentially insert guanosine. May play a role in hypermutation of immunoglobulin genes. Forms a Schiff base with 5'-deoxyribose phosphate at abasic sites, but may not have lyase activity.
Specifications
POLI | |
Polyclonal | |
Unconjugated | |
Poli | |
DNA polymerase iota; eta2; Poli; polymerase (DNA directed) iota; polymerase (DNA directed), iota; polymerase (DNA) iota; polymerase (DNA-directed), iota; RAD 30B; RAD30; RAD30 homolog B; RAD30B; RAD3OB | |
Rabbit | |
50 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot, Immunocytochemistry | |
Conc. Not Determined | |
Whole serum with 0.02% sodium azide | |
Q6R3M4, Q9UNA4 | |
Poli | |
A 15-mer peptide in the C-terminal region of human DNA Polymerase Iota, conjugated to KLH. | |
RUO | |
11201, 26447 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction