Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Policeman Spatula
SDPSCI_ED

Catalog No. S15679 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Length (Metric):
9.5 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
S15679 9.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S15679 Supplier United Scientific™ Supplier No. 81411
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Plastic design is a safe alternative to glass.

  • A multi-purpose tool made from polypropylene can be used as a stirrer or spatula.
  • Measures 9.5 in. long with a diameter of 0.25 in.
  • Pack of 12

Specifications

Quantity 12/Pk.
Blade Material Polypropylene
Handle Material Polypropylene
Material Polypropylene
Handle Type Round
Length (Metric) 9.5 in.
Autoclavable Autoclavable
Shape Flat End
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.