Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Poliovirus 3 Antibody (45 D5), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP105102F
Description
Poliovirus 3 Monoclonal antibody specifically detects Poliovirus 3 in Virus samples. It is validated for ELISASpecifications
Poliovirus 3 | |
Monoclonal | |
FITC | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Virus | |
Purified |
ELISA | |
45 D5 | |
ELISA | |
PV-3 | |
Poliovirus type 3 (inactivated) | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction