Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Poliovirus Antibody, FITC, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB10062836
Description
Poliovirus Polyclonal specifically detects Poliovirus in Virus samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
Poliovirus | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
Enterovirus C, poliomyelitis | |
Pool of purified poliovirus (types 1-3) | |
0.5 mL | |
Primary | |
Virus | |
Purified |
Immunocytochemistry, Immunofluorescence | |
FITC | |
PBS with 0.05% Sodium Azide | |
Goat | |
IgG purified | |
RUO | |
Specific for poliovirus. The will recognize poliovirus types 1, 2 and 3. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction