Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poliovirus receptor Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501735680 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5680 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5680 Supplier Proteintech Group Inc Supplier No. 274861AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PVR is a member of Ig-superfamily with 3 Ig-domains in arrangement V-C-C. Four mRNAs are produced from single gene for PVR, (mapped to human chromosome band 19q13.1-13.2). Two of the mRNAs (called H20A and H20B or alpha and kappa,) encode cell surface molecules of about 43kDa and 45kDa polypeptide backbone, respectively. Two other splice variants, beta and gamma encode secreted molecules of about 44kDa. The cytoplasmic tails are short (aa 35-50) and are rich in serine phosphorylated residues. MW of PVR is.

Specifications

Antigen Poliovirus receptor
Applications Western Blot
Classification Polyclonal
Concentration 0.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene PVR
Gene Accession No. P15151
Gene Alias CD155, HVED, Necl 5, NECL5, Nectin like protein 5, Poliovirus receptor, PVR, PVS, TAGE4
Gene Symbols PVR
Host Species Rabbit
Immunogen Poliovirus receptor Fusion Protein Ag26657
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5817
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.