Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Poliovirus Type 1-3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA173124
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA173124 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA173124 Supplier Invitrogen™ Supplier No. PA173124
Add to Cart
Add to Cart

Goat Polyclonal Antibody

The PA1-73124 antibody reacts with Poliovirus types 1-3. PA1-73124 has been successfully used in immunofluorescence applications. The PA1-73124 antibody was raised against a mixture of purified virus (types 1-3).

Poliovirus is a single-stranded RNA virus that belongs to the Picornavirus family, characterized by their small RNA genome (7.2bk to 8.5 kb). Poliovirus is a enterovirus which primarily infects the gastroentestinal assiciated lymphoid tissue (GALT) and can eventually spread to the central nervous system, causing paralysis. Mortality, though very rare, is usually due to respiratory failure by paralysis of the intercostal muscles and diaphragm. There are three separate types of the virus: Type 1, Type 2 and Type 3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Poliovirus Type 1-3
Applications Immunocytochemistry
Classification Polyclonal
Concentration 5.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias PV-1; PV-2; PV-3
Host Species Goat
Immunogen Mixture of purified virus (types 1-3).
Purification Method IgG Fraction
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.