Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POLL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104270
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104270 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104270 Supplier Invitrogen™ Supplier No. PA5104270
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total DNA pol lambda.

The protein encoded by this gene functions as a cofactor for the stimulation of transcriptional elongation by HIV-1 Tat, which binds to the HIV-1 promoter through Tat-TAR interaction. This protein may also serve as a dual-function factor to couple transcription and splicing and to facilitate their reciprocal activation. Alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POLL
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Poll
Gene Accession No. Q5RKI3, Q9QXE2, Q9UGP5
Gene Alias 1110003P06Rik; AV007317; BETAN; DNA polymerase beta-2; DNA polymerase beta-N; DNA polymerase kappa; DNA polymerase lambda; DNA-directed DNA polymerase lambda; EC 2.7.7.7; FLJ46002; OTTHUMP00000020323; Pol beta2; Pol Lambda; Polk; POLKAPPA; POL-KAPPA; Poll; polymerase (DNA directed), lambda; polymerase (DNA) lambda; RP11-529I10.1
Gene Symbols Poll
Host Species Rabbit
Immunogen A synthesized peptide derived from human POLL(Accession Q9UGP5), corresponding to amino acid residues V493-C543.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27343, 361767, 56626
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.