Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pollen allergen Phl p 1 Antibody (Clone 25), PerCP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP281276PCP
Description
Pollen allergen Phl p 1 Monoclonal antibody specifically detects Pollen allergen Phl p 1 in Plant samples. It is validated for Western Blot, ELISA, Dot BlotSpecifications
| Pollen allergen Phl p 1 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Plant | |
| Purified |
| Western Blot, ELISA, Dot Blot | |
| Clone 25 | |
| Western Blot, ELISA, Dot Blot | |
| Allergen Phl p I, grass pollen allergen | |
| Recombinant Phl p 1, a grass pollen allergen. | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction