Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POLM Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566220
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566220 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566220 Supplier Invitrogen™ Supplier No. PA566220
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ITHPKEGQEAG LLPRVMCRLQ DQGLILYHQH QHSCCESPTR LAQQSHMDAF ERSFCIFRLP QPPGAAVGGS TRPCPSWKAV RVDLVVAPVS QFP Highest antigen sequence identity to the following orthologs - mouse 66%, rat 67%.

Gap-filling polymerase involved in repair of DNA double-strand breaks by non-homologous end joining (NHEJ). Participates in immunoglobulin (Ig) light chain gene rearrangement in V(D)J recombination.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POLM
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene POLM
Gene Accession No. Q9NP87
Gene Alias AI462166; B230309I03Rik; DNA polymerase mu; DNA-directed DNA polymerase mu; DNA-directed DNA/RNA polymerase mu; Pol iota; pol Mu; Polm; polmu; polymerase (DNA directed), mu; polymerase (DNA) mu; polymerase (DNA-directed), mu; Tdt-N; Terminal transferase
Gene Symbols POLM
Host Species Rabbit
Immunogen Recombinant Human POLM. Recombinant protein control fragment (Product #RP-106901).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27434
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.