Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POLN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565161
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565161 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565161 Supplier Invitrogen™ Supplier No. PA565161
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FSTLTSQWKDV PVEQVTHADR EQTKKVVYAV VYGAGKERLA ACLGVPIQEA AQFLESFLQK YKKIKDFARA AIAQCHQTGC VVSIMGRRR Highest antigen sequence identity to the following orthologs - mouse 78%, rat 78%.

This gene encodes a DNA polymerase type-A family member. The encoded protein plays a role in DNA repair and homologous recombination. This gene shares its 5' exons with some transcripts from overlapping GeneID: 79441, which encodes an augmentin-like protein complex subunit. [provided by RefSeq, Dec 2014].
TRUSTED_SUSTAINABILITY

Specifications

Antigen POLN
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene POLN
Gene Accession No. Q7Z5Q5
Gene Alias DNA polymerase N; DNA polymerase nu; DNA polymerase POL4P; DNA-directed DNA polymerase nu; POL4P; POLN; polymerase (DNA directed) nu; polymerase (DNA) nu
Gene Symbols POLN
Host Species Rabbit
Immunogen Recombinant Human POLN. Recombinant protein control fragment (Product #RP-105910).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 353497
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.