Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Poloxamer 407, NF, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
2.5 kg
12 kg
25 kg
45 kg
Packaging:
Fiber Drum
Poly Drum
Poly Pail
Description
Spectrum™ Chemical Poloxamer 407, NF is a national formulary approved surfactant that finds utility because of its amphiphilic structure. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
pH | 5.0 to 7.5 |
CAS Max % | 1 |
Quantity | 45 kg |
Grade | NF |
Packaging | Poly Drum |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction