Learn More
Invitrogen™ POLQ Polyclonal Antibody

Description
The antibody detects endogenous levels of total DNA Polymerase theta protein.
Specifications
Specifications
| Antigen | POLQ |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | POLQ |
| Gene Accession No. | Q8CGS6 |
| Gene Alias | A430110D14Rik; Chaos1; Chromosome aberrations occurring spontaneously protein 1; DNA polymerase eta; DNA polymerase theta; DNA polymerase theta; LOW QUALITY PROTEIN: DNA polymerase theta; EC 2.7.7.7; POLH; Polq; polymerase (DNA directed), theta; polymerase (DNA) theta; PRO0327 |
| Gene Symbols | POLQ |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.