Learn More
POLR1A/RPA194 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304846A

Description
The recommended shelf life for this product is 1 year from date of receipt.
RNA polymerases transcribe nuclear genes for ribosomal RNA, thus representing ribosomal biogenesis. RNA polymerase I (Pol I) is located in the nucleolus and transcribes class I genes, which code for large ribosomal RNA. Different subunits of the Pol I transcription machinery are targets of various physiological stimuli, which suggests that multiple signaling pathways are involved in carrying out Pol I transcription. RPA40 and RPA16 are subunits of Pol I that associate with each other at an early stage of RNA polymerase I assembly. RPA40 is essential for the function and integrity of the complex and is also an essential subunit of RNA polymerase III (Pol III). RPA40, RPA16 and RPA135 encode the three subunits of RNA polymerase I, respectively. RPA194 is the largest subunit of RNA Pol I and is not a component of Pol II and Pol III.Specifications
POLR1A/RPA194 | |
Polyclonal | |
Unconjugated | |
POLR1A | |
DNA-directed RNA polymerase I subunit RPA1, RNA polymerase I subunit A1, A190, DNA-directed RNA polymerase I largest subunit, DNA-directed RNA polymerase I subunit A, RNA polymerase I 194 kDa subunit, RPA194, POLR1A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25885 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
O95602 | |
POLR1A | |
Between 1425 and 1475 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.