Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POLR1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588309
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588309 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588309 Supplier Invitrogen™ Supplier No. PA588309
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IANFIPFSDH NQSPRNMYQC QMGKQTMGFP LLTYQDRSDN KLYRLQTPQS PLVRPSMYDY YDMDNYPIGT NAIVAVISYT GYDMEDAMIV NKASWERGFA HGSVYKSEFI DLSEKIKQGD SSLVFGIKPG DPRVLQKLDD DGLPFIGAKL QYGDPYYSYL NLNTGESFVM YYKSKENCVV DNIKVCSNDT GSGKFKCVCI TMRVPRNPTI GDKFASRHGQ KGILSRLWPA E; Positive Samples: HeLa, HT-29, 293T, Mouse spleen; Cellular Location: Nucleus, nucleolus.

Eukaryotic RNA polymerase I (pol I) is responsible for the transcription of ribosomal RNA (rRNA) genes and production of rRNA, the primary component of ribosomes. Pol I is a multisubunit enzyme composed of 6 to 14 polypeptides, depending on the species. Most of the mass of the pol I complex derives from the 2 largest subunits, Rpa1 and Rpa2 in yeast. POLR1B is homologous to Rpa2 (Seither and Grummt, 1996).
TRUSTED_SUSTAINABILITY

Specifications

Antigen POLR1B
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.65 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Polr1b
Gene Accession No. P70700, Q9H9Y6
Gene Alias 128kDa; A127; D630020H17Rik; DNA-directed RNA polymerase I 135 kDa polypeptide; DNA-directed RNA polymerase I 135kDa polypeptide; DNA-directed RNA polymerase I subunit RPA2; Polr1b; polymerase (RNA) I polypeptide B; polymerase (RNA) I polypeptide B, 128kDa; polymerase (RNA) I subunit B; RNA polymerase 1-2; RNA polymerase 1-2, 128 kDa subunit; RNA polymerase I (127 kDa subunit); RNA polymerase I polypeptide B; RNA polymerase I subunit 2; RNA polymerase I subunit B; RPA116; RPA135; Rpa2; Rpo1-2
Gene Symbols Polr1b
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 670-900 of human POLR1B (NP_0618872).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20017, 84172
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.