Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POLR1C Monoclonal Antibody (2A5-A5)
GREENER_CHOICE

Catalog No. PIMA520761
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA520761 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA520761 Supplier Invitrogen™ Supplier No. MA520761
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MAASQAVEEM RSRVVLGEFG VRNVHTTDFP GNYSGYDDAW DQDRFEKNFR VDVVHMDENS LEFDMVGIDA AIANAFRRIL LAEVPTMAVE KVLVYNNTSI VQDEILAHRL GLIPIHADPR LFEYRNQGDE EGTEIDTLQF RLQVRCTRNP HAAKDSSDPN ELYVNHKVYT RHMTWIPLGN QADLFPEGTI RPVHDDILIA QLRPGQEIDL LMHCVKGIGK DHAKFSPVAT ASYRLLPDIT LLEPVEGEAA EELSRCFSPG VIEVQEVQGK KVARVANPRL DTFSREIFRN EKLKKVVRLA RVRDHYIFSV ESTGVLPPDV LVSEAIKVLM GKCRRFLDEL DAVQMD.

RNA polymerases transcribe nuclear genes for ribosomal RNA, thus representing ribosomal biogenesis. RNA polymerase I (Pol I) is located in the nucleolus and transcribes class I genes, which code for large ribosomal RNA. Different subunits of the Pol I transcription machinery are targets of various physiological stimuli, which suggests that multiple signaling pathways are involved in carrying out Pol I transcription. RPA16, RPA40 and RPA135 are subunits of Pol I that associate with each other at an early stage of RNA Pol I assembly. RPA40 is essential for the function and integrity of the complex and is also an essential subunit of RNA polymerase III (Pol III).
TRUSTED_SUSTAINABILITY

Specifications

Antigen POLR1C
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 2A5-A5
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene POLR1C
Gene Accession No. O15160
Gene Alias 40kDa; AA409007; AA959927; Ac2-127; AC40; AL024089; DNA-directed RNA polymerase I subunit C; DNA-directed RNA polymerases I and III 40 kDa polypeptide; DNA-directed RNA polymerases I and III subuni RPAC1-like protein; DNA-directed RNA polymerases I and III subunit RPAC1; HLD11; Polr1c; POLR1E; polymerase (RNA) I polypeptide C; polymerase (RNA) I polypeptide C, 30kDa; polymerase (RNA) I subunit C; RNA polymerase 1-1; RNA polymerase 1-1 (40 kDa subunit); RNA polymerase I 140 kDa subunit; RNA polymerase I and III subunit C; RNA polymerase I subunit; RNA polymerase I subunit C; RNA polymerase subunit I; RNA polymerases I and III subunit AC1; RPA39; RPA40; RPA5; RPAC1; RPC40; Rpo1-1; TCS3; Unknown (protein for MGC:134521); zgc:65973
Gene Symbols POLR1C
Host Species Mouse
Immunogen POLR1C (AAH08863, 1 a.a. ~ 346 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9533
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.