Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POLR1C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111008
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111008 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111008 Supplier Invitrogen™ Supplier No. PA5111008
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NNTSIVQDEI LAHRLGLIPI HADPRLFEYR NQGDEEGTEI DTLQFRLQVR CTRNPHAAKD SSDPNELYVN HKVYTRHMTW IPLGNQADLF PEGTIRPVHD Highest antigen sequence indentity to the following orthologs: Mouse - 95%, Rat - 93%.

RNA polymerases transcribe nuclear genes for ribosomal RNA, thus representing ribosomal biogenesis. RNA polymerase I (Pol I) is located in the nucleolus and transcribes class I genes, which code for large ribosomal RNA. Different subunits of the Pol I transcription machinery are targets of various physiological stimuli, which suggests that multiple signaling pathways are involved in carrying out Pol I transcription. RPA16, RPA40 and RPA135 are subunits of Pol I that associate with each other at an early stage of RNA Pol I assembly. RPA40 is essential for the function and integrity of the complex and is also an essential subunit of RNA polymerase III (Pol III).
TRUSTED_SUSTAINABILITY

Specifications

Antigen POLR1C
Applications ChIP Assay, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene POLR1C
Gene Accession No. O15160
Gene Alias 40kDa; AA409007; AA959927; Ac2-127; AC40; AL024089; DNA-directed RNA polymerase I subunit C; DNA-directed RNA polymerases I and III 40 kDa polypeptide; DNA-directed RNA polymerases I and III subuni RPAC1-like protein; DNA-directed RNA polymerases I and III subunit RPAC1; HLD11; Polr1c; POLR1E; polymerase (RNA) I polypeptide C; polymerase (RNA) I polypeptide C, 30kDa; polymerase (RNA) I subunit C; RNA polymerase 1-1; RNA polymerase 1-1 (40 kDa subunit); RNA polymerase I 140 kDa subunit; RNA polymerase I and III subunit C; RNA polymerase I subunit; RNA polymerase I subunit C; RNA polymerase subunit I; RNA polymerases I and III subunit AC1; RPA39; RPA40; RPA5; RPAC1; RPC40; Rpo1-1; TCS3; Unknown (protein for MGC:134521); zgc:65973
Gene Symbols POLR1C
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human POLR1C. Recombinant protein control fragment (Product #RP-94949).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9533
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.