Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POLR1D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121170
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121170 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121170 Supplier Invitrogen™ Supplier No. PA5121170
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HeLa, MCF7, A-549, SW480, Jurkat. The target is usually found in the following locations: Nucleus. Immunogen sequence: MEEDQELERK ISGLKTSMAE GERKTALEMV QAAGTDRHCV TFVLHEEDHT LGNSLRYMIM KNPEVEFCGY TTTHPSESKI NLRIQTRGTL PAVEPFQRGL NELMNVCQHV LDKFEASIKD YKDQKASRNE STF.

The protein encoded by this gene is a component of the RNA polymerase I and RNA polymerase III complexes, which function in the synthesis of ribosomal RNA precursors and small RNAs, respectively. Mutations in this gene are a cause of Treacher Collins syndrome (TCS), a craniofacial development disorder. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POLR1D
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4.71 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene POLR1D
Gene Accession No. P0DPB6
Gene Alias 1110003G10Rik; AC19; DNA-directed RNA polymerase I 16 kDa polypeptide; DNA-directed RNA polymerase I subunit D; DNA-directed RNA polymerases I and III subunit RPAC2; hRPA19; POLR1C; Polr1d; polymerase (RNA) I polypeptide D; polymerase (RNA) I polypeptide D, 16kDa; polymerase (RNA) I subunit D; Protein POLR1D, isoform 2; RNA polymerase 1-3; RNA polymerase I 16 kDa subunit; RNA polymerase I subunit D; RNA polymerases I and III subunit AC2; RPA16; RPA9; RPAC2; RPC16; Rpo1-3; TCS2
Gene Symbols POLR1D
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-133 of human POLR1D (NP_057056.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51082
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.