Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ POLR1D Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558742
Description
Immunogen sequence: EAKRGKTRAE TMGPMGWMKC PLASTNKRFL INTIKNTLPS HKEQDHEQKE GDKEPAKSQA QKEENPKKHR SHPYKHSFRA RGSASYSPPR KRSSQDKYEK RS Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 80%.
The protein encoded by this gene is a component of the RNA polymerase I and RNA polymerase III complexes, which function in the synthesis of ribosomal RNA precursors and small RNAs, respectively. Mutations in this gene are a cause of Treacher Collins syndrome (TCS), a craniofacial development disorder. Alternative splicing results in multiple transcript variants.
Specifications
| POLR1D | |
| Polyclonal | |
| Unconjugated | |
| POLR1D | |
| 1110003G10Rik; AC19; DNA-directed RNA polymerase I 16 kDa polypeptide; DNA-directed RNA polymerase I subunit D; DNA-directed RNA polymerases I and III subunit RPAC2; hRPA19; POLR1C; Polr1d; polymerase (RNA) I polypeptide D; polymerase (RNA) I polypeptide D, 16kDa; polymerase (RNA) I subunit D; Protein POLR1D, isoform 2; RNA polymerase 1-3; RNA polymerase I 16 kDa subunit; RNA polymerase I subunit D; RNA polymerases I and III subunit AC2; RPA16; RPA9; RPAC2; RPC16; Rpo1-3; TCS2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 51082 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P0DPB6 | |
| POLR1D | |
| Recombinant protein corresponding to Human POLR1D. Recombinant protein control fragment (Product #RP-98005). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction