Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POLR1E/PRAF1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501568809
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-8809 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-8809 Supplier Bethyl Laboratories, Inc Supplier No. A304835AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

DNA-dependent RNA polymerase catalyzes the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates. Component of RNA polymerase I which synthesizes ribosomal RNA precursors. Appears to be involved in the formation of the initiation complex at the promoter by mediating the interaction between Pol I and UBTF/UBF.

Specifications

Antigen POLR1E/PRAF1
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene POLR1E
Gene Accession No. Q9GZS1
Gene Alias PAF53, PRAF1, RP11-405L18.3, DNA-directed RNA polymerase I subunit E, DNA-directed RNA polymerase I subunit RPA49, RNA polymerase I associated factor 53, RNA polymerase I subunit A49, RNA polymerase I-associated factor 1, RNA polymerase I-associated factor 53, polymerase (RNA) I associated
Gene Symbols POLR1E
Host Species Rabbit
Immunogen Between 431 and 481
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64425
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.