Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POLR2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117034
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117034 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117034 Supplier Invitrogen™ Supplier No. PA5117034
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: SH-SY5Y, A375, HepG2, BT-474, Mouse brain, Mouse spleen, Mouse ovary, Rat ovary Immunogen sequence: MYDADEDMQY DEDDDEITPD LWQEACWIVI SSYFDEKGLV RQQLDSFDEF IQMSVQRIVE DAPPIDLQAE AQHASGEVEE PPRYLLKFEQ IYLSKPTHWE RDGAPSPMMP NEARLRNLTY SAPLYVDITK TVIKEGEEQL QTQHQKTFIG KIPIMLRSTY CLLNGLTDRD LCELNECPLD PGGYFIINGS EKVLIAQEKM ATNTVYVFAK KDSKYAYTGE CRSCLENSSR PTSTIWVSML ARGGQGAKKS AIGQRIVATL.

This gene encodes the second largest subunit of RNA polymerase II, the polymerase responsible for synthesizing messenger RNA in eukaryotes. This subunit, in combination with at least two other polymerase subunits, forms a structure within the polymerase that maintains contact in the active site of the enzyme between the DNA template and the newly synthesized RNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POLR2B
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.62 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene POLR2B
Gene Accession No. P30876, Q8CFI7
Gene Alias DNA directed RNA polymerase II polypeptide B; DNA-directed RNA polymerase II 140 kDa polypeptide; DNA-directed RNA polymerase II subunit B; DNA-directed RNA polymerase II subunit RPB2; hRPB140; hs hRPB140; hsRPB2; Pol2rb; POLR2B; polymerase (RNA) II (DNA directed) polypeptide B; polymerase (RNA) II (DNA directed) polypeptide B, 140kDa; polymerase (RNA) II subunit B; RNA polymerase II second largest subunit; RNA polymerase II subunit 2; RNA polymerase II subunit B; RNA polymerase II subunit B2; rpb 2; Rpb140; Rpb2
Gene Symbols POLR2B
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 231329, 289561, 5431
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.