Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POLR2B Polyclonal Antibody, Active Motif™
SDP

Rabbit Polyclonal Antibody

$156.65 - $620.58

Specifications

Antigen POLR2B
Concentration Lot-Specific
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-199-3495
SDP
Encompass
View Documents
Active Motif
61558
10 μL
Each for $156.65
Only null left
Add to Cart
 
50-199-3494
SDP
Encompass
View Documents
Active Motif
61557
100 μL
Each for $620.58
Only null left
Add to Cart
 
Description

Description

This gene encodes the second largest subunit of RNA polymerase II, the polymerase responsible for synthesizing messenger RNA in eukaryotes. This subunit, in combination with at least two other polymerase subunits, forms a structure within the polymerase that maintains contact in the active site of the enzyme between the DNA template and the newly synthesized RNA.
Specifications

Specifications

POLR2B
Immunoprecipitation, Western Blot
Unconjugated
Rabbit
Human
P30876
5431
This antibody was raised against a peptide within the N-terminal region of human POLR2B.
Primary
-20°C, Avoid Freeze/Thaw Cycles
POLR2B
Lot-Specific
Polyclonal
Liquid
RUO
PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
DNA directed RNA polymerase II polypeptide B; DNA-directed RNA polymerase II 140 kDa polypeptide; DNA-directed RNA polymerase II subunit B; DNA-directed RNA polymerase II subunit RPB2; hRPB140; hs hRPB140; hsRPB2; Pol2rb; POLR2B; polymerase (RNA) II (DNA directed) polypeptide B; polymerase (RNA) II (DNA directed) polypeptide B, 140kDa; polymerase (RNA) II subunit B; RNA polymerase II second largest subunit; RNA polymerase II subunit 2; RNA polymerase II subunit B; RNA polymerase II subunit B2; Rpb140; Rpb2
POLR2B
IgG
Antigen affinity chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.