Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POLR2C Antibody, Novus Biologicals™
SDP

Catalog No. NBP180817 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP180817 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP180817 Supplier Novus Biologicals Supplier No. NBP180817
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

POLR2C Polyclonal specifically detects POLR2C in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen POLR2C
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 ug/ml, Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. P19387
Gene Alias DNA-directed RNA polymerase II 33 kDa polypeptide, DNA-directed RNA polymerase II subunit C, DNA-directed RNA polymerase II subunit RPB3, hRPB33, hsRPB3, polymerase (RNA) II (DNA directed) polypeptide C (33kD), polymerase (RNA) II (DNA directed) polypeptide C, 33kDa, RNA polymerase II subunit 3, RNA polymerase II subunit B3, RPB3, RPB31, RPB33
Gene Symbols POLR2C
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:ALRHTVYPKPEEWPKSEYSELDEDESQAPYDPNGKPERFYYNVESCGSLRPETIVLSALSGLKKKLSDLQTQLSHEIQ
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Repair, DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 5432
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.