Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POLR2C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559745
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559745 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559745 Supplier Invitrogen™ Supplier No. PA559745
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VEFTLDVRCN EDQTRHVTSR DLISNSPRVI PVTSRNRDND PNDYVEQDDI LIVKLRKGQE LRLRAYAKKG FGKEHAKWNP TAGVAFEYDP D Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

DNA-dependent RNA polymerase catalyzes the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates. Component of RNA polymerase II which synthesizes mRNA precursors and many functional non-coding RNAs. Pol II is the central component of the basal RNA polymerase II transcription machinery. It is composed of mobile elements that move relative to each other. RPB3 is part of the core element with the central large cleft and the clamp element that moves to open and close the cleft.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POLR2C
Applications ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene POLR2C
Gene Accession No. P19387, P97760
Gene Alias 33kDa; A-152E5.7; DNA-directed RNA polymerase II 33 kDa polypeptide; DNA-directed RNA polymerase II subunit C; DNA-directed RNA polymerase II subunit RPB3; hRPB33; hsRPB3; mRBP31; Polr2c; polymerase (RNA) II (DNA directed) polypeptide C; polymerase (RNA) II (DNA directed) polypeptide C, 33kDa; polymerase (RNA) II subunit C; polymerase II polypeptide C; RNA polymerase II 3; RNA polymerase II subunit 3; RNA polymerase II subunit B3; RNA polymerase II subunit C; RPB3; RPB31; RPB33; Rpo2-3
Gene Symbols POLR2C
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human POLR2C. Recombinant protein control fragment (Product #RP-98622).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20021, 361365, 5432
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.