Learn More
Invitrogen™ RBP4 Monoclonal Antibody (RB42)

Description
Human plasma RBP4. All MAbs recognize free RBP4 and RBP4 in complex with transthyretin. All MAbs react with apo- as well as holo-RBP4.
Specifications
Specifications
| Antigen | RBP4 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | RB42 |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | RBP4 |
| Gene Accession No. | P02753 |
| Gene Alias | MCOPCB10; Plasma retinol binding protein 4 (PRBP); Plasma retinol-binding protein; Plasma retinol-binding protein(1-176); Plasma retinol-binding protein(1-179); Plasma retinol-binding protein(1-181); Plasma retinol-binding protein(1-182); PRBP; PRO2222; RBP; Rbp4; Rbp-4; RBPA; RDCCAS; retinol; retinol binding; retinol binding protein 4; Retinol binding protein 4 interstitial; retinol binding protein 4, cellular; retinol binding protein 4, plasma; retinol-binding protein 4; retinol-binding protein 4, interstitial |
| Gene Symbols | RBP4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.