Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POLR2D Polyclonal Antibody

Catalog No. PIPA535954
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA535954 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA535954 Supplier Invitrogen™ Supplier No. PA535954
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, HeLa, HepG2. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (96%), Xenopus laevis (93%), Chicken (97%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes the fourth largest subunit of RNA polymerase II, the polymerase responsible for synthesizing messenger RNA in eukaryotes. In yeast, this polymerase subunit is associated with the polymerase under suboptimal growth conditions and may have a stress protective role. A sequence for a ribosomal pseudogene is contained within the 3' untranslated region of the transcript from this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POLR2D
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.01 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol; pH 7
Gene Polr2d
Gene Accession No. O15514
Gene Alias 2310002G05Rik; 2610028L19Rik; DNA-directed RNA polymerase II 16 kDa polypeptide; DNA-directed RNA polymerase II subunit D; DNA-directed RNA polymerase II subunit RPB4; HSRBP4; HSRPB4; plasma; POLR2D; polymerase (RNA) II (DNA directed) polypeptide D; polymerase (RNA) II subunit D; RBP4; RNA polymerase II 16 kDa subunit; RNA polymerase II subunit B4; RNA polymerase II subunit D; RNA polymerase II subunit hsRBP4; RPB16
Gene Symbols Polr2d
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human POLR2D. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5433
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.