Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ POLR2D Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA576635
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
This gene encodes the fourth largest subunit of RNA polymerase II, the polymerase responsible for synthesizing messenger RNA in eukaryotes. In yeast, this polymerase subunit is associated with the polymerase under suboptimal growth conditions and may have a stress protective role. A sequence for a ribosomal pseudogene is contained within the 3' untranslated region of the transcript from this gene.
Specifications
POLR2D | |
Polyclonal | |
Unconjugated | |
Polr2d | |
2310002G05Rik; 2610028L19Rik; DNA-directed RNA polymerase II 16 kDa polypeptide; DNA-directed RNA polymerase II subunit D; DNA-directed RNA polymerase II subunit RPB4; HSRBP4; HSRPB4; plasma; POLR2D; polymerase (RNA) II (DNA directed) polypeptide D; polymerase (RNA) II subunit D; RBP4; RNA polymerase II 16 kDa subunit; RNA polymerase II subunit B4; RNA polymerase II subunit D; RNA polymerase II subunit hsRBP4; RPB16 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5433, 69241 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunoprecipitation, Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
O15514, Q9D7M8 | |
Polr2d | |
Recombinant full length Human POLR2D. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction