Learn More
POLR2E Mouse anti-Canine, Human, Mouse, Rat, Clone: OTI3B5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF502548

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes the fifth largest subunit of RNA polymerase II, the polymerase responsible for synthesizing messenger RNA in eukaryotes. This subunit is shared by the other two DNA-directed RNA polymerases and is present in two-fold molar excess over the other polymerase subunits. An interaction between this subunit and a hepatitis virus transactivating protein has been demonstrated, suggesting that interaction between transcriptional activators and the polymerase can occur through this subunit. A pseudogene is located on chromosome 11.Specifications
POLR2E | |
Monoclonal | |
Unconjugated | |
POLR2E | |
RPABC1, RPB5, XAP4, hRPB25, hsRPB5 | |
Mouse | |
Affinity Chromatography | |
RUO | |
5434, 611859, 66420, 690966 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI3B5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
B0BNE2, P19388, Q80UW8 | |
Polr2e | |
Full length human recombit protein of human POLR2E produced in HEK293T cell. | |
100 μg | |
Primary | |
Canine, Human, Mouse, Rat | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.