Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POLR2E Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564389
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564389 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564389 Supplier Invitrogen™ Supplier No. PA564389
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RTDLTVLVAH NDDPTDQMFV FFPEEPKVGI KTIKVYCQRM QEENITRALI VVQQGMTPSA KQSLVDMAPK YILEQFLQQ Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

This gene encodes the fifth largest subunit of RNA polymerase II, the polymerase responsible for synthesizing messenger RNA in eukaryotes. This subunit is shared by the other two DNA-directed RNA polymerases and is present in two-fold molar excess over the other polymerase subunits. An interaction between this subunit and a hepatitis virus transactivating protein has been demonstrated, suggesting that interaction between transcriptional activators and the polymerase can occur through this subunit. A pseudogene is located on chromosome 11.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POLR2E
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene POLR2E
Gene Accession No. P19388
Gene Alias 2410021N14Rik; 25kDa; AW208866; DNA directed RNA polymerase II 23 kda polypeptide; DNA-directed RNA polymerase II 23 kDa polypeptide; DNA-directed RNA polymerase II subunit E; DNA-directed RNA polymerase subunit RPABC1; DNA-directed RNA polymerases I, II, and III subunit RPABC1; hRPB25; hsRPB5; Polr2e; polymerase (RNA) II (DNA directed) polypeptide E; polymerase (RNA) II (DNA directed) polypeptide E, 25kDa; polymerase (RNA) II subunit E; RNA polymerase II subunit E; RNA polymerase II, I and III subunit E; RNA polymerases I, II, and III subunit ABC1; RPABC1; RPB5; RPB5 homolog; XAP4
Gene Symbols POLR2E
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human POLR2E. Recombinant protein control fragment (Product #RP-104001).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5434
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.