Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POLR2F Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103803
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103803 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103803 Supplier Invitrogen™ Supplier No. PA5103803
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RPAB2.

DNA-dependent RNA polymerases catalyze the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates. Common component of RNA polymerases I, II, and III which synthesize ribosomal RNA precursors, mRNA precursors and many functional non-coding RNAs, and small RNAs, such as 5S rRNA and tRNAs, respectively. Pol II is the central component of the basal RNA polymerase II transcription machinery. Pols are composed of mobile elements that move relative to each other. In Pol II, POLR2F/RPB6 is part of the clamp element and together with parts of RPB1 and RPB2 forms a pocket to which the RPB4-RPB7 subcomplex binds.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POLR2F
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene POLR2F
Gene Accession No. O88828, P61218, P61219
Gene Alias 1810060D16Rik; DNA-directed RNA polymerase II subunit F; DNA-directed RNA polymerases I, II, and III 14.4 kDa polypeptide; DNA-directed RNA polymerases I, II, and III subunit RPABC2; HRBP14.4; hsRPB6; Polr2f; POLRF; polymerase (RNA) II (DNA directed) polypeptide F; polymerase (RNA) II subunit F; polymerase II; RNA Pol II; RNA Polymerase II subunit 14.4 kD; RNA polymerase II subunit F; RNA polymerases I, II, and III subunit ABC2; RPABC14.4; RPABC2; RPB14.4; RPB6; RPB6 homolog; RPC15
Gene Symbols POLR2F
Host Species Rabbit
Immunogen A synthesized peptide derived from human POLR2F(Accession P61218), corresponding to amino acid residues M1-L39.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5435, 69833, 83503
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.