Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POLR2G Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA521984
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA521984 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA521984 Supplier Invitrogen™ Supplier No. PA521984
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (98%), Sheep (99%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes the seventh largest subunit of RNA polymerase II, the polymerase responsible for synthesizing messenger RNA in eukaryotes. The protein functions in transcription initiation, and is also thought to help stabilize transcribing polyermase molecules during elongation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POLR2G
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene POLR2G
Gene Accession No. P62487, P62488
Gene Alias 2410046K11Rik; A230108L04Rik; C76415; DNA directed RNA polymerase II 19 kda polypeptide; DNA-directed RNA polymerase II 19 kDa polypeptide; DNA-directed RNA polymerase II subunit G; DNA-directed RNA polymerase II subunit RPB7; h hsRPB7; hRPB19; hs; hsRPB7; MGC138367; MGC138369; POLR2G; polymerase (RNA) II (DNA directed) polypeptide G; polymerase (RNA) II subunit G; RBP7; RNA polymerase II 19 kDa subunit; RNA polymerase II seventh subunit; RNA polymerase II subunit B7; RNA polymerase II subunit G; RPB19; RPB7; Rpo2-7l
Gene Symbols POLR2G
Host Species Rabbit
Immunogen Full length human POLR2G Recombinant protein.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5436, 67710
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.