Learn More
Invitrogen™ POLR2G Polyclonal Antibody

Description
Antibody detects endogenous levels of total RPB7.
Specifications
Specifications
| Antigen | POLR2G |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | POLR2G |
| Gene Accession No. | P62487, P62488, P62489 |
| Gene Alias | 2410046K11Rik; A230108L04Rik; C76415; DNA directed RNA polymerase II 19 kda polypeptide; DNA-directed RNA polymerase II 19 kDa polypeptide; DNA-directed RNA polymerase II subunit G; DNA-directed RNA polymerase II subunit RPB7; h hsRPB7; hRPB19; hs; hsRPB7; MGC138367; MGC138369; POLR2G; polymerase (RNA) II (DNA directed) polypeptide G; polymerase (RNA) II subunit G; RBP7; RNA polymerase II 19 kDa subunit; RNA polymerase II seventh subunit; RNA polymerase II subunit B7; RNA polymerase II subunit G; RPB19; RPB7; Rpo2-7l |
| Gene Symbols | POLR2G |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.