Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ POLR2G Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA519171
Description
This antibody is predicted to react with mouse, ovine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1,000.
This gene encodes the seventh largest subunit of RNA polymerase II, the polymerase responsible for synthesizing messenger RNA in eukaryotes. The protein functions in transcription initiation, and is also thought to help stabilize transcribing polyermase molecules during elongation.
Specifications
POLR2G | |
Polyclonal | |
Unconjugated | |
POLR2G | |
2410046K11Rik; A230108L04Rik; C76415; DNA directed RNA polymerase II 19 kda polypeptide; DNA-directed RNA polymerase II 19 kDa polypeptide; DNA-directed RNA polymerase II subunit G; DNA-directed RNA polymerase II subunit RPB7; h hsRPB7; hRPB19; hs; hsRPB7; MGC138367; MGC138369; POLR2G; polymerase (RNA) II (DNA directed) polypeptide G; polymerase (RNA) II subunit G; RBP7; RNA polymerase II 19 kDa subunit; RNA polymerase II seventh subunit; RNA polymerase II subunit B7; RNA polymerase II subunit G; RPB19; RPB7; Rpo2-7l | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
5436 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P62487 | |
POLR2G | |
Synthetic peptide sequence (EFDPNSNPPCYK) corresponding to the internal amino acids of POLR2G (aa 118-130). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction