Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POLR2H Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120207
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120207 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120207 Supplier Invitrogen™ Supplier No. PA5120207
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: SW480, MCF7, HL-60, 293T, Mouse spleen, Mouse thymus. The target is usually found in the following locations: Nucleus, nucleolus. Immunogen sequence: MAGILFEDIF DVKDIDPEGK KFDRVSRLHC ESESFKMDLI LDVNIQIYPV DLGDKFRLVI ASTLYEDGTL DDGEYNPTDD RPSRADQFEY VMYGKVYRIE GDETSTEAAT RLSAYVSYGG LLMRLQGDAN NLHGFEVDSR VYLLMKKLAF.

Eukaryotes produce 3 distinct classes of RNA polymerase, Pol I, II and III. Each polymerase is responsible for the synthesis of a different class of RNA. RNA polymerase I (Pol I) transcribes the rRNA (ribosomal RNA) genes for the precursor of the 28S, 18S, and 5. 8S molecules of the ribosome. RNA polymerase II transcribes protein-encoding genes into mRNA (messenger RNA) and snRNA (small nuclear RNA) genes into snRNAs that influence the processing of other classes of RNA. RNA polymerase III (Pol III) transcribes the 5S rRNA genes and all of the tRNA (transfer RNA) genes. Each class of RNA polymerase is assembled from 9 to 15 different polypeptides. The RPB6 and RPB8 subunits are shared by all 3 RNA polymerases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POLR2H
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 2.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene POLR2H
Gene Accession No. P52434, Q923G2
Gene Alias ABC3; DNA-directed RNA polymerase II subunit H; DNA-directed RNA polymerases I, II, and III 17.1 kDa polypeptide; DNA-directed RNA polymerases I, II, and III subunit RPABC3; hRPB8; hsRPB8; POLR2H; polymerase (RNA) II (DNA directed) polypeptide H; polymerase (RNA) II subunit H; RGD1561203; RNA polymerase II subunit H; RNA polymerases I, II, and III subunit ABC3; RPABC3; RPB17; RPB8; RPB8 homolog
Gene Symbols POLR2H
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human POLR2H (NP_006223.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 245841, 5437
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.