Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POLR2J Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560587
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560587 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560587 Supplier Invitrogen™ Supplier No. PA560587
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DPQVLFAGYK VPHPLEHKII IRVQTTPDYS PQEAFTNAIT DLISELSLLE ERFR Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

DNA-dependent RNA polymerase catalyzes the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates. Component of RNA polymerase II which synthesizes mRNA precursors and many functional non-coding RNAs. Pol II is the central component of the basal RNA polymerase II transcription machinery. It is composed of mobile elements that move relative to each other. RPB11 is part of the core element with the central large cleft.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POLR2J
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene POLR2J
Gene Accession No. P52435
Gene Alias 14.5kDa; DNA-directed RNA polymerase II 13.3 kDa polypeptide; DNA-directed RNA polymerase II subunit J; DNA-directed RNA polymerase II subunit J-1; DNA-directed RNA polymerase II subunit RPB11; DNA-directed RNA polymerase II subunit RPB11-a; hRPB14; Polr2i; POLR2J; POLR2J1; polymerase (RNA) II (DNA directed) polypeptide I; polymerase (RNA) II (DNA directed) polypeptide J; polymerase (RNA) II (DNA directed) polypeptide J, 13.3kDa; polymerase (RNA) II subunit J; RNA polymerase II 13.3 kDa subunit; RNA polymerase II 4 (14 kDa subunit); RNA polymerase II subunit B11; RNA polymerase II subunit B11-a; RNA polymerase II subunit J; RNA polymerase II subunit RPB14; RPB11; RPB11A; RPB11m; RPB14; Rpo2-4
Gene Symbols POLR2J
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human POLR2J. Recombinant protein control fragment (Product #RP-104365).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5439
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.