Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POLR2J Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548675
Description
Predicted to react with bovine based on sequence homology.
DNA-dependent RNA polymerase catalyzes the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates. Component of RNA polymerase II which synthesizes mRNA precursors and many functional non-coding RNAs. Pol II is the central component of the basal RNA polymerase II transcription machinery. It is composed of mobile elements that move relative to each other. RPB11 is part of the core element with the central large cleft.
Specifications
POLR2J | |
Polyclonal | |
PBS with 0.09% sodium azide | |
P52435 | |
POLR2J | |
KLH conjugated synthetic peptide between 89-117 amino acids from the C-terminal region of human POLR2J. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
POLR2J | |
DNA-directed RNA polymerase II subunit RPB11-a, RNA polymerase II subunit B11-a, RPB11a, DNA-directed RNA polymerase II subunit J-1, RNA polymerase II 133 kDa subunit, POLR2J, POLR2J1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5439 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction