Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POLR2J Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501730925 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-0925 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-0925 Supplier Proteintech Group Inc Supplier No. 164031AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Three genes (POLR2 J1, POLR2 J2, and POLR2 J3) form a cluster of total length of 214 530 bp in the genetic locus 7q22.1 on the long arm of chromosome 7 (contig NT_007933). The fourth gene (POLR2 J4, 31 040 bp) was localized in the cytogenetic locus 7p13 of the short arm of chromosome 7 (contig NT_007819). An analysis enabled us to refine dissimilar experimental data on the mapping of the hRPB11 subunit gene on chromosome 7. In particular, the presence of three sites of its localization according to data on hybridization with fluorescent-labeled probes (the FISH method) was explained. It was established that, upon the expression of the four human POLR2 J genes, at least 14 types of mature mRNAs encoding somewhat differing hRPB11 isoforms can be synthesized. Eleven of these mRNAs were revealed (as full-length copies or clearly identifiable fragments) in the available databases of expressed sequence tags and cDNAs. The most probable scheme of origination of the multiple genes of the POLR2 J family as a result of three consecutive segmented duplications increasing in size was proposed and substantiated. On the basis of the scheme, some assumptions on the pathways of evolution of separate human genes and the mechanisms of generation of protein diversity in higher eukaryotes were made

Specifications

Antigen POLR2J
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.15 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene POLR2J
Gene Accession No. O08740, P52435
Gene Alias hRPB14, POLR2J, POLR2J1, RPB11, RPB11A, RPB11m
Gene Symbols Polr2j
Host Species Rabbit
Immunogen POLR2J Fusion Protein Ag9520
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20022, 288588, 5439
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.