Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POLR2J2 Antibody (OTI3G4), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273520F
Description
POLR2J2 Monoclonal specifically detects POLR2J2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
POLR2J2 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
POLR2J2 | |
Full length recombinant protein of human POLR2J2 (NP_116581) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG2a |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI3G4 | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
DNA directed RNA polymerase II polypeptide J-related, DNA-directed RNA polymerase II subunit RPB11-b1, HRPB11B, MGC105050, MGC54043, polymerase (RNA) II (DNA directed) polypeptide J2, RNA polymerase II subunit B11-b1, RPB11, RPB11b1DNA-directed RNA polymerase II subunit J2 | |
Mouse | |
Affinity Purified | |
DNA replication Transcription Translation and Splicing | |
246721.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction