Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POLR2J2 Antibody (OTI3G4), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273520MFV610
Description
POLR2J2 Monoclonal antibody specifically detects POLR2J2 in Human samples. It is validated for Flow CytometrySpecifications
POLR2J2 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
OTI3G4 | |
Flow Cytometry | |
DNA directed RNA polymerase II polypeptide J-related, DNA-directed RNA polymerase II subunit RPB11-b1, HRPB11B, MGC105050, MGC54043, polymerase (RNA) II (DNA directed) polypeptide J2, RNA polymerase II subunit B11-b1, RPB11, RPB11b1DNA-directed RNA polymerase II subunit J2 | |
Full length human recombinant protein of human POLR2J2 (NP_116581) produced in HEK293T cell. | |
0.1 mL | |
DNA replication Transcription Translation and Splicing | |
246721 | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction