Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POLR2J2 Monoclonal Antibody (OTI3G4)
GREENER_CHOICE

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA525426

Catalog No. PIMA525426


Only null left
Add to Cart

Description

Description

POLR2J2 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

This gene is a member of the RNA polymerase II subunit 11 gene family, which includes three genes in a cluster on chromosome 7q22.1 and a pseudogene on chromosome 7p13. The founding member of this family, DNA directed RNA polymerase II polypeptide J, has been shown to encode a subunit of RNA polymerase II, the polymerase responsible for synthesizing messenger RNA in eukaryotes. This locus produces multiple, alternatively spliced transcripts that potentially express isoforms with distinct C-termini compared to DNA directed RNA polymerase II polypeptide J. Most or all variants are spliced to include additional non-coding exons at the 3' end which makes them candidates for nonsense-mediated decay (NMD). Consequently, it is not known if this locus expresses a protein or proteins in vivo.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

POLR2J2
Monoclonal
0.72 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9GZM3
POLR2J2
Full length human recombinant protein of POLR2J2 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG2a
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
OTI3G4
Unconjugated
POLR2J2
DNA directed RNA polymerase II polypeptide J-related; DNA-directed RNA polymerase II subunit J2; DNA-directed RNA polymerase II subunit RPB11-b1; HRPB11B; POLR2J2; polymerase (RNA) II (DNA directed) polypeptide J2; polymerase (RNA) II subunit J2; RNA polymerase II subunit B11-b1; RNA polymerase II subunit J2; RPB11b1
Mouse
Affinity Chromatography
RUO
246721
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.